Learn More
Hach Company Boiler/Condens Conduct Sens
Catalog No. 5000207733 Shop All Hach Company Products
Encompass
Analog Boiler/Condensate Conductivity Sensor-Cell Constant 5 0-Stainless Steel Body-PP HDNominal Cell Constant (K) 5 0 3/4 inch male NPT threaded 316 stainless steel body Polypropylene Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode Generic ID
Catalog No. 50-002-07733 Supplier Hach Company Supplier No. 3444D8B
Analog Boiler/Condensate Conductivity Sensor-Cell Constant 5 0-Stainless Steel Body-PP HDNominal Cell Constant (K) 5 0 3/4 inch male NPT threaded 316 stainless steel body Polypropylene Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode Generic ID