Learn More
Fisher Science Education™ Boiling Chips
SCI_ED
Catalog No. S25201 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
12/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25201 100 g
S25201C 12/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25201
Add to Cart
Edge
Add to Cart
Helps prevent bumping.
These inert non-volatile granules resist most chemical action except those of concentrated sulfuric acids and strong alkalies. And, they won't contaminate distillate.
This product is for educational use - no C of A is available.
Specifications
Type Boiling Chips
Material Marble
Quantity 100 g