Learn More
Innovating Science™ Boiling Chips, Marble
SCI_ED
Catalog No. S93141 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
S93141 500 g
1 options
Catalog No. S93141 Supplier Innovating Science™ Supplier No. ADCC0060500G
Add to Cart
Add to Cart
For use with distillations.
  • Chemically inert marble chips, CaCO3
  • Promote gentle boiling and minimize bumping and superheating
  • 500 g bottle
Specifications
Type Boiling Stone
Material Marble
For Use With (Equipment) Chemware Glassware
Quantity 500 g