Learn More
Thomas Scientific 133B Carborundum #12 Granules, 500g
SDP
Supplier:  Thomas Scientific 1590D33
Encompass
Hard, bluish black, iridescent silicon carbide granule. No reduction characteristics. Oxidizes slowly at 800°C (1472°F). Twelve mesh granule size. Specific gravity 3.20. 5 year industry leading warranty In any laboratory procedure involving the boiling of a liquid, inert and insoluble, Hengar Granules effectively prevent super-heating with its attendant dangerous explosive vaporization commonly known as "bumping". A few Hengar Granules in any boiling liquid permit a steady copious stream of bubbles to escape without the spasmodic splatter. Evaporation or distillation proceeds quietly and smoothly. Sharper fractions result. Flask breakage is reduced and watching time is minimized. Froth is spread out evenly and does not climb the neck of the flask. Especially useful in boiling soap solutions and evaporating ethereal extracts of fat. With Hengar Granules in the vessel, any gases formed are extracted from the liquid quickly and completely. Insoluble, Hengar Granules cann
Catalog No. NC9063828