Learn More
METTLER TOLEDO 3P INDUSTRIAL BP/CP/SMP outer Capillaries
Catalog No. NC0190946
Encompass
High-quality glass capillaries that are perfect for boiling point, cloud point and slip melting point analyses with the Melting Point Excellence instruments.
Catalog No. NC0190946 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 18572
High-quality glass capillaries that are perfect for boiling point, cloud point and slip melting point analyses with the Melting Point Excellence instruments.