Learn More
Accuflow Systems Polytetrafluoroethylene Boiling Stones | 450 GR
Catalog No. NC2327705
Encompass_Preferred
Polytetrafluoroethylene (PTFE) boiling stones | Chemical resistant, Ideal for boiling organic liquids, Minimizes superheating | 450g
Catalog No. NC2327705 Supplier Accuflow Systems Supplier No. D1069103
Polytetrafluoroethylene (PTFE) boiling stones | Chemical resistant, Ideal for boiling organic liquids, Minimizes superheating | 450g