Learn More
Aviva Systems Biology Corp Bola1 Antibody - N-terminal region (ARP37374_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37374_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Bola1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the n terminal region of mouse Bola1. Alternate names: 1810037G04RikApplicationWB. Species reactivity: Human, Mouse, Rabbit, Rat
Catalog No. 50-284-621