Learn More
Aviva Systems Biology Corp Bola2 Antibody - N-terminal region (ARP37542_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37542_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Bola2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the n terminal region of mouse Bola2. Alternate names: 1110025L05Rik, BolA-2ApplicationWB. Species reactivity: Cow, Goat, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-653