Learn More
ABclonal Technology BOLA3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15985500UL
Encompass_Preferred
This gene encodes a protein that plays an essential role in the production of iron-sulfur (Fe-S) clusters for the normal maturation of lipoate-containing 2-oxoacid dehydrogenases, and for the assembly of the mitochondrial respiratory chain complexes. Mutation in this gene has been associated with multiple mitochondrial dysfunctions syndrome-2. Two alternatively spliced transcript variants encoding different isoforms with distinct subcellular localization have been reported for this gene (PMID21944046).
Catalog No. 50-001-08207