Learn More
Cayman Chemical Boldenone Undecylenate, Purity: ≥98%, CAS: 13103-34-9, 5 mg
SDP
Catalog No. 502986193
Encompass_Preferred
An analytical reference standard categorized as an anabolic androgenic steroid; has been used to enhance physical performance in racehorses and athletes; intended for research and forensic applications
Catalog No. 50-298-6193 Supplier Cayman Chemical Supplier No. 216325 MG
An analytical reference standard categorized as an anabolic androgenic steroid; has been used to enhance physical performance in racehorses and athletes; intended for research and forensic applications