Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boldine, MilliporeSigma™ Supelco™

Catalog No. 111014249 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4249 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4249 Supplier MilliporeSigma Supelco Supplier No. B39165G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boldine is an aporphine alkaloid isolated from Boldo tree and other plant species. It is a free radical scavenger with anti-inflammatory activity while exhibiting other pharmacological effects such as antidiabetic, antiplatelet aggregation, antipyretic, antinociceptive, antiatherogenic, hepatoprotective and endothelium-protective activity. Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 476-70-0
Physical Form Neat
Linear Formula C19H21 NO4
MDL Number MFCD00135040
Synonym 2,9-Dihydroxy-1,10-dimethoxyaporphine
Type Analytical Standard
Quantity 5 g
Formula Weight 327.37
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.