Learn More
Aviva Systems Biology Corp BOLL Antibody - N-terminal region (ARP41065_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41065_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BOLL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BOLL. Alternate names: BOULEApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57707