Learn More
MSC Bolle SAFETY Atom Goggle Visor Shield, ANSI Z87.1-2010, 1 ea
Catalog No. 5000793369
Encompass
This Goggle Visor Shield is specifically designed for use with Bolle Atom Goggles, providing additional face protection. It functions as an Atom face shield to enhance user safety.
  • Compatible with Atom goggles
  • Functions as a safety glass dispenser and goggle visor shield
  • Provides goggle side shields
  • Features a blue color
  • Meets ANSI Z87.1-2010 standards
Catalog No. 50-007-93369 Supplier MSC Supplier No. 72591779
This Goggle Visor Shield is specifically designed for use with Bolle Atom Goggles, providing additional face protection. It functions as an Atom face shield to enhance user safety.
  • Compatible with Atom goggles
  • Functions as a safety glass dispenser and goggle visor shield
  • Provides goggle side shields
  • Features a blue color
  • Meets ANSI Z87.1-2010 standards