Learn More
MSC Bolle Safety Glasses, Anti-Fog, Scratch-Resistant, Polycarbonate Bronze Lenses, 1 ea
Catalog No. 5000793368
Encompass
The Cobra safety glasses provide panoramic vision and a wrap-around fit for optimal eye protection. Featuring a frameless design, these versatile glasses offer both anti-fog and scratch-resistant properties.
  • Anti-fog coatings improve visibility in hot and humid conditions
  • Scratch-resistant lenses enhance durability
  • Panoramic visual field with 180° vision
  • Wrap-around fit without visual interference
  • Pivoting temples allow for adjustment and improved protection
  • Option to swap temples for an adjustable strap and foam seal
  • ANSI Z87.1-2010 compliant
Catalog No. 50-007-93368 Supplier MSC Supplier No. 72591530
The Cobra safety glasses provide panoramic vision and a wrap-around fit for optimal eye protection. Featuring a frameless design, these versatile glasses offer both anti-fog and scratch-resistant properties.
  • Anti-fog coatings improve visibility in hot and humid conditions
  • Scratch-resistant lenses enhance durability
  • Panoramic visual field with 180° vision
  • Wrap-around fit without visual interference
  • Pivoting temples allow for adjustment and improved protection
  • Option to swap temples for an adjustable strap and foam seal
  • ANSI Z87.1-2010 compliant