Learn More
MSC Bolle Safety Glasses, Anti-Fog and Scratch-Resistant, Polycarbonate, Full-Framed, 1 ea
Catalog No. 5000685221
Encompass
These safety glasses are designed with clear, anti-fog and scratch-resistant polycarbonate lenses within a full-framed, wrap-around design. They feature a gray nylon frame with adjustable gray temples and provide UV protection, complying with ANSI Z87.1-2010 standards.
  • Anti-fog and scratch-resistant lenses
  • Polycarbonate clear lenses
  • Full-framed, wrap-around design
  • Gray nylon frame with adjustable temples
  • Provides UV protection
  • Meets ANSI Z87.1-2010 standards
  • Features adjustable non-slip bridge type
Catalog No. 50-006-85221 Supplier MSC Supplier No. 76907492
These safety glasses are designed with clear, anti-fog and scratch-resistant polycarbonate lenses within a full-framed, wrap-around design. They feature a gray nylon frame with adjustable gray temples and provide UV protection, complying with ANSI Z87.1-2010 standards.
  • Anti-fog and scratch-resistant lenses
  • Polycarbonate clear lenses
  • Full-framed, wrap-around design
  • Gray nylon frame with adjustable temples
  • Provides UV protection
  • Meets ANSI Z87.1-2010 standards
  • Features adjustable non-slip bridge type