Learn More
MSC Bolle Safety Goggles, Chemical Splash, Anti-Fog, Clear Polycarbonate, 1 ea.
Catalog No. 5000793932
Encompass
These safety goggles offer reliable eye protection against chemical splash, dust, and impact. They feature clear polycarbonate lenses with anti-fog and scratch-resistant coatings, ensuring clear vision in challenging conditions. Designed for universal fit, they can be worn over prescription glasses.
  • Indirect ventilation for enhanced comfort
  • Provides protection against impact, dust, and chemical splash
  • Clear polycarbonate lenses with anti-fog and scratch-resistant features
  • Frame constructed from clear PVC
  • Universal size designed to fit over prescription glasses
  • Complies with ANSI Z87.1-2010 standards
  • Includes a durable nylon polyester strap
Catalog No. 50-007-93932 Supplier MSC Supplier No. 72591704
These safety goggles offer reliable eye protection against chemical splash, dust, and impact. They feature clear polycarbonate lenses with anti-fog and scratch-resistant coatings, ensuring clear vision in challenging conditions. Designed for universal fit, they can be worn over prescription glasses.
  • Indirect ventilation for enhanced comfort
  • Provides protection against impact, dust, and chemical splash
  • Clear polycarbonate lenses with anti-fog and scratch-resistant features
  • Frame constructed from clear PVC
  • Universal size designed to fit over prescription glasses
  • Complies with ANSI Z87.1-2010 standards
  • Includes a durable nylon polyester strap