Learn More
MSC Bolle Safety Goggles, Pilot Neo, Brown Polycarbonate, Anti-Fog, 1 ea
Catalog No. 5000766797
Encompass
Pilot Neo Safety Goggles are designed for industrial applications, providing protection against dust and splash hazards with a comfortable, over-the-glasses fit.
  • Anti-fog and scratch-resistant polycarbonate lenses
  • Indirect ventilation for enhanced comfort
  • Brown lenses for improved visibility in various conditions
  • Meets ANSI Z87+ D3 D4 standards
  • Equipped with an adjustable neoprene strap
Catalog No. 50-007-66797 Supplier MSC Supplier No. 62136809
Pilot Neo Safety Goggles are designed for industrial applications, providing protection against dust and splash hazards with a comfortable, over-the-glasses fit.
  • Anti-fog and scratch-resistant polycarbonate lenses
  • Indirect ventilation for enhanced comfort
  • Brown lenses for improved visibility in various conditions
  • Meets ANSI Z87+ D3 D4 standards
  • Equipped with an adjustable neoprene strap