Learn More
MSC Bolle Safety Safety Glasses, Anti-Fog and Scratch-Resistant, 1 ea
Catalog No. 5000685413
Encompass
Bolle Safety Prism Series safety glasses provide eye protection with smoke lenses and a full-framed design. These glasses feature anti-fog and scratch-resistant coatings for clear vision and are adapted for use in the food-processing industry.
  • Lightweight frame
  • No metal parts
  • Anti-fog coatings improve visibility under hot and humid conditions
  • UV protection
  • ANSI Z87.1-2015 compliant
Catalog No. 50-006-85413 Supplier MSC Supplier No. 72591316
Bolle Safety Prism Series safety glasses provide eye protection with smoke lenses and a full-framed design. These glasses feature anti-fog and scratch-resistant coatings for clear vision and are adapted for use in the food-processing industry.
  • Lightweight frame
  • No metal parts
  • Anti-fog coatings improve visibility under hot and humid conditions
  • UV protection
  • ANSI Z87.1-2015 compliant