Learn More
MSC Bolle SAFETY Safety Glasses, Anti-Fog, Scratch-Resistant, Polycarbonate, 1 ea
Catalog No. 5000765506
Encompass
These bolle SAFETY safety glasses feature anti-fog and scratch-resistant polycarbonate clear lenses with a frameless design, black frame, and non-adjustable temples. They are part of the Rush+ series and meet ANSI Z87.1-2010 standards.
  • Anti-fog and scratch-resistant lenses
  • Polycarbonate clear lenses
  • Frameless, wrap-around design
  • Black frame and temples
  • Small size
  • Non-adjustable temples
  • UV protection
  • Meets ANSI Z87.1-2010 standards
  • Panoramic field of vision
  • Anti-fog coatings improve visibility under hot and humid conditions
Catalog No. 50-007-65506 Supplier MSC Supplier No. 37279999
These bolle SAFETY safety glasses feature anti-fog and scratch-resistant polycarbonate clear lenses with a frameless design, black frame, and non-adjustable temples. They are part of the Rush+ series and meet ANSI Z87.1-2010 standards.
  • Anti-fog and scratch-resistant lenses
  • Polycarbonate clear lenses
  • Frameless, wrap-around design
  • Black frame and temples
  • Small size
  • Non-adjustable temples
  • UV protection
  • Meets ANSI Z87.1-2010 standards
  • Panoramic field of vision
  • Anti-fog coatings improve visibility under hot and humid conditions