Learn More
MSC Bolle SAFETY Safety Glasses, Anti-Fog, Scratch-Resistant, Polycarbonate, 1 ea
Catalog No. 5000685098
Encompass
These safety glasses are designed with anti-fog and scratch-resistant polycarbonate lenses, providing clear vision and protection in various environments. Their full-framed, wrap-around design ensures comprehensive coverage.
  • Anti-fog coatings improve visibility under hot and humid conditions
  • Scratch-resistant lens coating
  • Full-framed design with a wrap-around frame
  • Clear polycarbonate lenses
  • Universal size for broad compatibility
  • Non-slip adjustable bridge for enhanced comfort
  • Provides UV protection
  • Flexible temples
  • Meets ANSI Z87.1-2010 standards
Catalog No. 50-006-85098 Supplier MSC Supplier No. 72591209
These safety glasses are designed with anti-fog and scratch-resistant polycarbonate lenses, providing clear vision and protection in various environments. Their full-framed, wrap-around design ensures comprehensive coverage.
  • Anti-fog coatings improve visibility under hot and humid conditions
  • Scratch-resistant lens coating
  • Full-framed design with a wrap-around frame
  • Clear polycarbonate lenses
  • Universal size for broad compatibility
  • Non-slip adjustable bridge for enhanced comfort
  • Provides UV protection
  • Flexible temples
  • Meets ANSI Z87.1-2010 standards