Learn More
MSC Bolle SAFETY Welding Helmet, Digital Controls, Polycarbonate, Multi-Shade, 1 ea
Catalog No. 5000674712
Encompass
This welding helmet features digital controls, a black polycarbonate construction, and auto-darkening lens shades from 5 and 8 to 13. It includes a fixed front window measuring 3.9300 in (100.00 mm) wide by 2.0800 in (53.00 mm) high, powered by both solar and battery sources, and meets ANSI Z87+ standards.
  • Blocking of the cell to clear state for grinding
  • Fine adjustment and progressive sensitivity
  • Auto recovery delay adjustment
  • Solar power
  • Low-battery indicator
  • Replaceable battery
  • Polycarbonate is a thermoplastic polymer, impact resistant and extremely tough
  • Low tolerance to scratches
Catalog No. 50-006-74712 Supplier MSC Supplier No. 58285081
This welding helmet features digital controls, a black polycarbonate construction, and auto-darkening lens shades from 5 and 8 to 13. It includes a fixed front window measuring 3.9300 in (100.00 mm) wide by 2.0800 in (53.00 mm) high, powered by both solar and battery sources, and meets ANSI Z87+ standards.
  • Blocking of the cell to clear state for grinding
  • Fine adjustment and progressive sensitivity
  • Auto recovery delay adjustment
  • Solar power
  • Low-battery indicator
  • Replaceable battery
  • Polycarbonate is a thermoplastic polymer, impact resistant and extremely tough
  • Low tolerance to scratches