Learn More
MSC Bolle Safety Welding Safety Glasses, Green Lens, Anti-Fog, 1 ea
Catalog No. 5000766904
Encompass
These full-framed safety glasses are designed to fit over prescription eyewear, featuring anti-fog and scratch-resistant polycarbonate green lenses. Engineered for welding applications, they provide a shade 5 protection and meet ANSI Z87.1-2010 standards, ensuring visibility and eye safety in hazardous environments.
  • Over-the-glasses (OTG) design
  • Anti-fog and scratch-resistant lens coating
  • Green lenses with welding shade 5
  • Universal size with adjustable temples
  • Provides 99.9% UVA/UVB protection
  • Complies with ANSI Z87.1-2010 and CSA-Z94.3-2007 standards
  • Polycarbonate lens and nylon frame material
  • Includes cord
Catalog No. 50-007-66904 Supplier MSC Supplier No. 72591365
These full-framed safety glasses are designed to fit over prescription eyewear, featuring anti-fog and scratch-resistant polycarbonate green lenses. Engineered for welding applications, they provide a shade 5 protection and meet ANSI Z87.1-2010 standards, ensuring visibility and eye safety in hazardous environments.
  • Over-the-glasses (OTG) design
  • Anti-fog and scratch-resistant lens coating
  • Green lenses with welding shade 5
  • Universal size with adjustable temples
  • Provides 99.9% UVA/UVB protection
  • Complies with ANSI Z87.1-2010 and CSA-Z94.3-2007 standards
  • Polycarbonate lens and nylon frame material
  • Includes cord