Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bolt™ Bis-Tris Welcome Pack, 8%

Catalog No. p-7224322
Encompass_Preferred
Change view
Click to view available options
Wells:
10-well
12-well
15-well
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells
NW0008A 10-well
NW0008B 12-well
NW0008C 15-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NW0008A Supplier Invitrogen™ Supplier No. NW0008A
Only null left
Add to Cart
Add to Cart

The Bolt Welcome Pack provides all of the necessary Bolt gels and buffers needed to begin using the Mini Gel Tank.

The Bolt Welcome Pack provides all of the necessary Bolt gels and buffers you will need to begin using the Mini Gel Tank. The Mini Gel Tank is compatible with Novex gels, NuPAGE gels, and Bolt Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt Bis-Tris Plus gels to be run in approximately 20 minutes.

The Welcome Pack contains:
• Mini Gel Tank (A25977)
• Bolt Bis-Tris Plus gels (2 boxes, 20 gels)
• Bolt MES Running Buffer, 20X (B0002)
• Bolt LDS Sample Buffer, 4X (B0007)
• Bolt Sample Reducing Agent, 10X (B0009)
• PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)

About the Mini Gel Tank
The Mini Gel Tank is compatible with Novex gels, NuPAGE gels, and Bolt Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt Bis-Tris Plus gels to be run in approximately 20 minutes. Run times may vary depending on buffer conditions and power supplies used.

About Bolt Bis-Tris Plus Gels
Bolt Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of your small- to medium-sized proteins under denaturing conditions. Bolt Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt gels to obtain optimal results for your day-to-day protein separation needs. Bolt Bis-Tris Plus gels come in four polyacrylamide concentrations: 8, 10, 12, and 4-12% gradient and multiple well formats. Bolt Bis-Tris gels feature wedge-shaped wells, which can easily load up to two times more sample volume allowing for detection of proteins in very dilute samples or visualization of low-abundance proteins.

For transferring your proteins to a membrane, we recommend using for Bolt Transfer Buffer (BT0006) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

Warranty and Services

Up to 12 months

TRUSTED_SUSTAINABILITY

Specifications

Gel Percentage 8%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Bis-Tris
Separation Range 3.5 to 180 kDa
Wells 10-well
Quantity 1 Welcome Pk. kit
Shipping Condition Approved for shipment at room temperature and wet ice
Product Line Bolt, WedgeWell
For Use With (Equipment) Mini Gel Tank
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.