Learn More
Gold Standard Diagnostics BOLT™ One-Plate ELISA + CLIA Analyzer
SDP
Catalog No. 23700161
Change view
Click to view available options
Type:
Automated Enzyme Immunoassay Analyzer
Catalog No. Type
23-700-161 Automated Enzyme Immunoassay Analyzer
1 options
This item is not returnable. View return policy
Catalog No. 23-700-161 Supplier Gold Standard Diagnostics Supplier No. 00500
Add to Cart
Add to Cart
This item is not returnable. View return policy
A one-plate ELISA+CLIA processor offering customizable options with lower throughput and a cost-effective design.
  • Compact one-plate analyzer supporting ELISA and CLIA assays
  • Shares core features of the ThunderBolt™ with flexible customization
  • Lower throughput design ideal for smaller labs or specific testing needs
  • Cost-effective system that reduces overall laboratory expenses
  • User-friendly interactive software for simplified operation
  • Manufactured by Gold Standard Diagnostics with proven reliability
Specifications
Type Automated Enzyme Immunoassay Analyzer
Capacity Up to 96 Samples
Wavelength 405, 450, 490, 550 and 630 nm
Electrical Requirements 100/265 V
Dimensions (L x W x H) 53.3 x 48.3 x 55.8 cm