Learn More
Grainger Boltless Shelving 18x84x48in AdOn
Catalog No. 502753544
Encompass_Preferred
The Grainger 4-Shelf Boltless Steel Shelving Unit is an industrial-grade freestanding shelving system made from steel with a powder-coated finish. It features adjustable shelves with interlocking posts and beams that assemble without tools or hardware. The shelves can support up to 800 lbs evenly distributed. The unit measures 36" W x 18" D x 74" H. Optional accessories include additional shelves, casters, bins and more. It's ideal for warehouses, garages, retail stores and other commercial/industrial settings that require sturdy shelving.
Catalog No. 50-275-3544 Supplier Grainger Supplier No. 14C691
The Grainger 4-Shelf Boltless Steel Shelving Unit is an industrial-grade freestanding shelving system made from steel with a powder-coated finish. It features adjustable shelves with interlocking posts and beams that assemble without tools or hardware. The shelves can support up to 800 lbs evenly distributed. The unit measures 36" W x 18" D x 74" H. Optional accessories include additional shelves, casters, bins and more. It's ideal for warehouses, garages, retail stores and other commercial/industrial settings that require sturdy shelving.