Learn More
Grainger Boltless Shelving 18x84x48in AdOn
Catalog No. 502751218
Encompass_Preferred
The Grainger 14L145 is a 14-inch diameter, 145 CFM, 115V, 1/3 HP exhaust fan. It features a galvanized steel housing with baked enamel finish, backward inclined aluminum blades, and permanently lubricated ball bearings. This exhaust fan has a maximum RPM of 1075 and operates at 68 dBA. It is suitable for exhausting air from commercial and industrial settings. The 14L145 can mount to ceilings or walls with included mounting brackets and plugs into a standard 115V outlet. This exhaust fan meets OSHA standard 1910.94(a)(4) for air movement.
Catalog No. 50-275-1218 Supplier Grainger Supplier No. 14L145
The Grainger 14L145 is a 14-inch diameter, 145 CFM, 115V, 1/3 HP exhaust fan. It features a galvanized steel housing with baked enamel finish, backward inclined aluminum blades, and permanently lubricated ball bearings. This exhaust fan has a maximum RPM of 1075 and operates at 68 dBA. It is suitable for exhausting air from commercial and industrial settings. The 14L145 can mount to ceilings or walls with included mounting brackets and plugs into a standard 115V outlet. This exhaust fan meets OSHA standard 1910.94(a)(4) for air movement.