Learn More
Grainger Boltless Shelving 18x84x60in Strtr
Catalog No. 502753413
Encompass_Preferred
The Grainger 14C701 is a 14 gauge, type 1 electrical extension cord that is 50 feet long. It has a yellow jacket and is designed for indoor and outdoor use. The cord has a grounded plug on one end and a grounded receptacle on the other end. The maximum amperage rating is 15 amps with 1875 watts at 125 volts AC. The cord can withstand temperatures from -31 to 140 degrees Fahrenheit. The jacket material is sunlight and oil resistant. This extension cord can be used for power tools, appliances, lighting, and more. It's UL listed and CSA certified.
Catalog No. 50-275-3413 Supplier Grainger Supplier No. 14C701
The Grainger 14C701 is a 14 gauge, type 1 electrical extension cord that is 50 feet long. It has a yellow jacket and is designed for indoor and outdoor use. The cord has a grounded plug on one end and a grounded receptacle on the other end. The maximum amperage rating is 15 amps with 1875 watts at 125 volts AC. The cord can withstand temperatures from -31 to 140 degrees Fahrenheit. The jacket material is sunlight and oil resistant. This extension cord can be used for power tools, appliances, lighting, and more. It's UL listed and CSA certified.