Learn More
Grainger Boltless Shelving 18x84x96in AdOn
Catalog No. 502753632
Encompass_Preferred
The Grainger 14C661 is a Westward brand 1/4-inch drive socket set. It contains 15 SAE sockets ranging from 1/4 inch to 9/16 inch and 15 metric sockets ranging from 4mm to 14mm. The chrome vanadium sockets have a satin finish and feature a stamped size on the socket for easy identification. This 108-piece set comes organized in a molded case for convenient storage and portability. The case has a hinged lid and socket rail to keep the sockets securely in place.
Catalog No. 50-275-3632 Supplier Grainger Supplier No. 14C661
The Grainger 14C661 is a Westward brand 1/4-inch drive socket set. It contains 15 SAE sockets ranging from 1/4 inch to 9/16 inch and 15 metric sockets ranging from 4mm to 14mm. The chrome vanadium sockets have a satin finish and feature a stamped size on the socket for easy identification. This 108-piece set comes organized in a molded case for convenient storage and portability. The case has a hinged lid and socket rail to keep the sockets securely in place.