Learn More
Grainger Boltless Shelving 24x84x60in AdOn
Catalog No. 502748839
Encompass_Preferred
The Boltless Shelving 48 in x 18 in x 72 in is a 4-shelf steel shelving unit. It has a weight capacity of 800 lbs per shelf and 3,200 lbs total. The shelves are adjustable in 1-inch increments. Assembly required using provided hardware. Measures 48" W x 18" D x 72" H overall. Features a boltless design for easy assembly. Shelves are constructed from steel with a black powder coated finish. Offered by Global Industrial Equipment through Grainger.
Catalog No. 50-274-8839 Supplier Grainger Supplier No. 14C643
The Boltless Shelving 48 in x 18 in x 72 in is a 4-shelf steel shelving unit. It has a weight capacity of 800 lbs per shelf and 3,200 lbs total. The shelves are adjustable in 1-inch increments. Assembly required using provided hardware. Measures 48" W x 18" D x 72" H overall. Features a boltless design for easy assembly. Shelves are constructed from steel with a black powder coated finish. Offered by Global Industrial Equipment through Grainger.