Learn More
Grainger Boltless Shelving 24x84x60in Strtr
Catalog No. 502752859
Encompass_Preferred
The Grainger 14C703 is a 14-inch diameter, 7-blade, cast aluminum fan blade suitable for exhaust and supply fans. It has a pitch of 3 inches and is balanced for quiet operation. The blade is unpainted and corrosion-resistant. It is designed for direct drive arrangements up to 1/2 HP and belt drive arrangements from 1/2 to 3/4 HP. The blade conforms to AMCA standard 99-0401-86. It can be used with Grainger's 4C546 motor for a complete fan assembly.
Catalog No. 50-275-2859 Supplier Grainger Supplier No. 14C703
The Grainger 14C703 is a 14-inch diameter, 7-blade, cast aluminum fan blade suitable for exhaust and supply fans. It has a pitch of 3 inches and is balanced for quiet operation. The blade is unpainted and corrosion-resistant. It is designed for direct drive arrangements up to 1/2 HP and belt drive arrangements from 1/2 to 3/4 HP. The blade conforms to AMCA standard 99-0401-86. It can be used with Grainger's 4C546 motor for a complete fan assembly.