Learn More
Grainger Boltless Shelving 30x84x60in AdOn
Catalog No. 502751375
Encompass_Preferred
The Hallowell Boltless Shelving is a steel shelving unit that can be assembled without tools or hardware. It has 4 adjustable shelves that can hold up to 800 lbs per shelf. The unit is 78" H x 36" W x 18" D. It is made from steel with a gray epoxy finish and can be used for industrial, retail, or office storage needs. The boltless design allows for quick assembly.
Catalog No. 50-275-1375 Supplier Grainger Supplier No. 14L159
The Hallowell Boltless Shelving is a steel shelving unit that can be assembled without tools or hardware. It has 4 adjustable shelves that can hold up to 800 lbs per shelf. The unit is 78" H x 36" W x 18" D. It is made from steel with a gray epoxy finish and can be used for industrial, retail, or office storage needs. The boltless design allows for quick assembly.