Learn More
Grainger Boltless Shelving 30x84x60in Strtr
Catalog No. 502751553
Encompass_Preferred
The Tennsco Boltless Shelving is a heavy duty steel shelving unit that can be assembled without tools or hardware. It has a weight capacity of 800 lbs per shelf. The unit has 4 adjustable shelves that can be spaced in 1" increments along the uprights. It measures 36"W x 18"D x 78"H. The shelves are constructed of 22 gauge steel and have rolled edges for safety. The uprights have holes punched every inch for adjustability. Assembly requires no nuts, bolts, clips or hardware. The finish is an attractive gray powder coat paint. This shelving unit provides sturdy, customizable storage for warehouses, garages, basements and more.
Catalog No. 50-275-1553 Supplier Grainger Supplier No. 14L158
The Tennsco Boltless Shelving is a heavy duty steel shelving unit that can be assembled without tools or hardware. It has a weight capacity of 800 lbs per shelf. The unit has 4 adjustable shelves that can be spaced in 1" increments along the uprights. It measures 36"W x 18"D x 78"H. The shelves are constructed of 22 gauge steel and have rolled edges for safety. The uprights have holes punched every inch for adjustability. Assembly requires no nuts, bolts, clips or hardware. The finish is an attractive gray powder coat paint. This shelving unit provides sturdy, customizable storage for warehouses, garages, basements and more.