Learn More
Grainger Boltless Shelving 30x84x72in AdOn
Catalog No. 502752686
Encompass_Preferred
The Edsal Boltless Shelving Unit is a 4-shelf steel unit offering versatile storage. Its boltless design allows for quick assembly without nuts or bolts. The 4 shelves adjust in 1-inch increments for customizable storage. Made from steel with a black epoxy finish, it measures 36" W x 18" D x 74" H and supports up to 800 lbs of evenly distributed weight. Ideal for garage, basement, or warehouse storage, its open design ensures easy access and visibility. This heavy-duty, industrial-strength unit is perfect for organizing storage spaces.
Catalog No. 50-275-2686 Supplier Grainger Supplier No. 14C716
The Edsal Boltless Shelving Unit is a 4-shelf steel unit offering versatile storage. Its boltless design allows for quick assembly without nuts or bolts. The 4 shelves adjust in 1-inch increments for customizable storage. Made from steel with a black epoxy finish, it measures 36" W x 18" D x 74" H and supports up to 800 lbs of evenly distributed weight. Ideal for garage, basement, or warehouse storage, its open design ensures easy access and visibility. This heavy-duty, industrial-strength unit is perfect for organizing storage spaces.