Learn More
Grainger Boltless Shelving 30x84x96in Strtr
Catalog No. 502751889
Encompass_Preferred
The Grainger 14C664 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA 48Y frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body. It has a C-face mounting style and measures approximately 9-3/4" long by 7-3/8" wide by 8-3/4" high. The 14C664 operates on single phase 115V power and draws 8.4 amps at full load.
Catalog No. 50-275-1889 Supplier Grainger Supplier No. 14C664
The Grainger 14C664 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA 48Y frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body. It has a C-face mounting style and measures approximately 9-3/4" long by 7-3/8" wide by 8-3/4" high. The 14C664 operates on single phase 115V power and draws 8.4 amps at full load.