Learn More
Grainger Boltless Shelving 36x84x48in Strtr
Catalog No. 502752117
Encompass_Preferred
The Grainger 14L150 is a Westward brand 1/4-inch drive click-type torque wrench. It has a torque range of 10 to 150 inch pounds. The wrench has a reversible ratchet head and comes with a storage case. The overall length is 15-1/2 inches. This torque wrench is designed for accurate tightening and can help prevent overtightening. It would be useful for automotive and machinery applications where proper torque values need to be maintained. The click-type mechanism provides both an audible click and tactile feedback when the preset torque value is reached.
Catalog No. 50-275-2117 Supplier Grainger Supplier No. 14L150
The Grainger 14L150 is a Westward brand 1/4-inch drive click-type torque wrench. It has a torque range of 10 to 150 inch pounds. The wrench has a reversible ratchet head and comes with a storage case. The overall length is 15-1/2 inches. This torque wrench is designed for accurate tightening and can help prevent overtightening. It would be useful for automotive and machinery applications where proper torque values need to be maintained. The click-type mechanism provides both an audible click and tactile feedback when the preset torque value is reached.