Learn More
Grainger Boltless Shelving 36x84x60in AdOn
Catalog No. 502750244
Encompass_Preferred
The Grainger 14L101 is a 14-inch diameter, 101-inch long, schedule 40 black steel pipe. It has a plain end construction and is made from ASTM A53 grade B ERW steel. This pipe has a wall thickness of 0.237 inches, weighs approximately 28.55 lbs per foot, and can withstand pressures up to 700 psi. The pipe's inside diameter measures 13.124 inches. It can be used for a variety of industrial piping applications.
Catalog No. 50-275-0244 Supplier Grainger Supplier No. 14L101
The Grainger 14L101 is a 14-inch diameter, 101-inch long, schedule 40 black steel pipe. It has a plain end construction and is made from ASTM A53 grade B ERW steel. This pipe has a wall thickness of 0.237 inches, weighs approximately 28.55 lbs per foot, and can withstand pressures up to 700 psi. The pipe's inside diameter measures 13.124 inches. It can be used for a variety of industrial piping applications.