Learn More
Grainger Boltless Shelving 36x84x60in AdOn
Catalog No. 502751646
Encompass_Preferred
The Boltonless Shelving unit by Tennsco is a heavy duty steel shelving unit that is 36"W x 84"H x 60"D. It has a weight capacity of 8000 lbs per shelf. The unit has 4 adjustable shelves that can be moved in 1" increments. Assembly required using nuts and bolts (included). Finish is grey powder coat paint. Shelves are constructed from 22 gauge steel and uprights from 14 gauge steel. Can be used for industrial, retail or office storage needs. Ships via truck; check website for freight shipping quote.
Catalog No. 50-275-1646 Supplier Grainger Supplier No. 14L161
The Boltonless Shelving unit by Tennsco is a heavy duty steel shelving unit that is 36"W x 84"H x 60"D. It has a weight capacity of 8000 lbs per shelf. The unit has 4 adjustable shelves that can be moved in 1" increments. Assembly required using nuts and bolts (included). Finish is grey powder coat paint. Shelves are constructed from 22 gauge steel and uprights from 14 gauge steel. Can be used for industrial, retail or office storage needs. Ships via truck; check website for freight shipping quote.