Learn More
Grainger Boltless Shelving 36x84x60in AdOn
Catalog No. 502752270
Encompass_Preferred
The Grainger 14C708 is a 14 gauge, type C, 7/8" wide, 8' long copper grounding braid that can be used for flexible ground connections. It is made in the USA out of oxygen-free high conductivity electrolytic tough-pitch copper for optimal conductivity and has a current capacity of 100 amps. This ground braid can be cut to length for custom applications. It meets UL, CSA, IEC, and NEC standards for copper grounding braids. The braid features tinned copper strands and is RoHS compliant.
Catalog No. 50-275-2270 Supplier Grainger Supplier No. 14C708
The Grainger 14C708 is a 14 gauge, type C, 7/8" wide, 8' long copper grounding braid that can be used for flexible ground connections. It is made in the USA out of oxygen-free high conductivity electrolytic tough-pitch copper for optimal conductivity and has a current capacity of 100 amps. This ground braid can be cut to length for custom applications. It meets UL, CSA, IEC, and NEC standards for copper grounding braids. The braid features tinned copper strands and is RoHS compliant.