Learn More
Grainger Boltless Shelving 36x84x60in AdOn
Catalog No. 502748761
Encompass_Preferred
The Tennsco Boltless Shelving is a heavy duty steel shelving unit that can be assembled without tools or hardware. It has a weight capacity of 800 lbs per shelf. The unit has 4 adjustable shelves that are 30" deep x 36" wide. The overall dimensions are 36"W x 18"D x 72"H. The shelves are made of 22 gauge steel and have rolled edges for safety. The posts have foot plates to protect floors. This shelving unit can be used in industrial, commercial, or residential settings for storage and organization. It comes in a gray finish to resist corrosion.
Catalog No. 50-274-8761 Supplier Grainger Supplier No. 14C647
The Tennsco Boltless Shelving is a heavy duty steel shelving unit that can be assembled without tools or hardware. It has a weight capacity of 800 lbs per shelf. The unit has 4 adjustable shelves that are 30" deep x 36" wide. The overall dimensions are 36"W x 18"D x 72"H. The shelves are made of 22 gauge steel and have rolled edges for safety. The posts have foot plates to protect floors. This shelving unit can be used in industrial, commercial, or residential settings for storage and organization. It comes in a gray finish to resist corrosion.