Learn More
Grainger Boltless Shelving 36x84x72in Strtr
Catalog No. 502751519
Encompass_Preferred
The Boltless Shelving Unit by Strtr is a 36"W x 84"H x 72"D steel shelving unit with 4 adjustable shelves. It has a weight capacity of 800 lbs per shelf and 3,200 lbs total. The unit assembles without tools using zinc-plated steel connectors and can be reconfigured. The shelves are constructed from steel with 3 ridges for strength and adjust in 1-1/2" increments on center posts. Optional casters can be added for mobility. This industrial-grade boltless shelving provides sturdy and customizable storage.
Catalog No. 50-275-1519 Supplier Grainger Supplier No. 14L170
The Boltless Shelving Unit by Strtr is a 36"W x 84"H x 72"D steel shelving unit with 4 adjustable shelves. It has a weight capacity of 800 lbs per shelf and 3,200 lbs total. The unit assembles without tools using zinc-plated steel connectors and can be reconfigured. The shelves are constructed from steel with 3 ridges for strength and adjust in 1-1/2" increments on center posts. Optional casters can be added for mobility. This industrial-grade boltless shelving provides sturdy and customizable storage.