Learn More
Grainger Boltless Shelving 36x84x96in AdOn
Catalog No. 502750139
Encompass_Preferred
The Grainger 14L121 is a 14-inch diameter, 121-tooth circular saw blade with a 5/8-inch arbor hole and a 0.062-inch kerf. It is designed for cutting wood and wood products. The blade has carbide teeth and is optimized for making crosscuts and miter cuts in softwood and hardwood. It can cut wood up to 3-1/2 inches thick. The blade should be run at speeds between 4000-6000 RPM for optimal performance. This circular saw blade is manufactured in the USA by Freud.
Catalog No. 50-275-0139 Supplier Grainger Supplier No. 14L121
The Grainger 14L121 is a 14-inch diameter, 121-tooth circular saw blade with a 5/8-inch arbor hole and a 0.062-inch kerf. It is designed for cutting wood and wood products. The blade has carbide teeth and is optimized for making crosscuts and miter cuts in softwood and hardwood. It can cut wood up to 3-1/2 inches thick. The blade should be run at speeds between 4000-6000 RPM for optimal performance. This circular saw blade is manufactured in the USA by Freud.