Learn More
Grainger Boltless Shelving 36x84x96in Strtr
Catalog No. 502753213
Encompass_Preferred
The Tennsco Boltless Shelving Unit is a heavy duty steel shelving system that provides adjustable storage space. It measures 36"W x 84"H x 96"D and has four adjustable shelves that can be moved in 1" increments. The unit has a weight capacity of 8000 lbs per shelf when evenly distributed. It features a boltless design for quick assembly and disassembly. The finish is an attractive gray powder coat paint that resists chipping and scratching. This shelving unit is ideal for warehouses, retail stores, garages and other commercial/industrial settings that require sturdy and customizable storage solutions.
Catalog No. 50-275-3213 Supplier Grainger Supplier No. 14L180
The Tennsco Boltless Shelving Unit is a heavy duty steel shelving system that provides adjustable storage space. It measures 36"W x 84"H x 96"D and has four adjustable shelves that can be moved in 1" increments. The unit has a weight capacity of 8000 lbs per shelf when evenly distributed. It features a boltless design for quick assembly and disassembly. The finish is an attractive gray powder coat paint that resists chipping and scratching. This shelving unit is ideal for warehouses, retail stores, garages and other commercial/industrial settings that require sturdy and customizable storage solutions.