Learn More
Grainger Boltless Shelving 48x84x72in AdOn
Catalog No. 502749932
Encompass_Preferred
The Tennsco Boltless Shelving is a heavy duty steel shelving unit that can be assembled without tools or hardware. It has a weight capacity of 800 lbs per shelf. The unit has 4 adjustable shelves that are 30" deep x 36" wide. The overall dimensions are 36"W x 18"D x 72"H. The shelves are made of 22 gauge steel and have rolled edges for safety. The posts have foot plates for stability. This shelving unit provides a sturdy storage solution for warehouses, garages, basements and more.
Catalog No. 50-274-9932 Supplier Grainger Supplier No. 14L173
The Tennsco Boltless Shelving is a heavy duty steel shelving unit that can be assembled without tools or hardware. It has a weight capacity of 800 lbs per shelf. The unit has 4 adjustable shelves that are 30" deep x 36" wide. The overall dimensions are 36"W x 18"D x 72"H. The shelves are made of 22 gauge steel and have rolled edges for safety. The posts have foot plates for stability. This shelving unit provides a sturdy storage solution for warehouses, garages, basements and more.