Learn More
MSC Value Collection Boltless Cross Beam, Steel, 96 in., 14 Gauge, 1 ea
Catalog No. 5000512450
Encompass
This boltless cross beam is designed for integration with boltless storage rack systems to provide robust support for organizational needs.
  • Constructed from durable 14-gauge steel
  • Finished with a powder coating for enhanced resilience
  • Designed for compatibility with boltless storage rack systems
  • Features a gray color finish
  • Measures 96 in. in height and 18 in. in width
Catalog No. 50-005-12450 Supplier MSC Supplier No. 41525346
This boltless cross beam is designed for integration with boltless storage rack systems to provide robust support for organizational needs.
  • Constructed from durable 14-gauge steel
  • Finished with a powder coating for enhanced resilience
  • Designed for compatibility with boltless storage rack systems
  • Features a gray color finish
  • Measures 96 in. in height and 18 in. in width