Learn More
Grainger Bolts Universal Fit
Catalog No. 502750981
Encompass_Preferred
The KISSLER Universal Fit Bolt is a grade 5 hex head bolt made from medium carbon alloy steel with a zinc yellow dichromate finish. It has a tensile strength of 120,000 psi. The bolt has a hex head with a washer face and is available in diameters ranging from 1/4"-3/4" in various lengths from 1" to 8". The bolt threads are UNC. This bolt can be used for general purpose fastening applications.
Catalog No. 50-275-0981 Supplier Grainger Supplier No. 14U362
The KISSLER Universal Fit Bolt is a grade 5 hex head bolt made from medium carbon alloy steel with a zinc yellow dichromate finish. It has a tensile strength of 120,000 psi. The bolt has a hex head with a washer face and is available in diameters ranging from 1/4"-3/4" in various lengths from 1" to 8". The bolt threads are UNC. This bolt can be used for general purpose fastening applications.