Learn More
Grainger Bolts Universal Fit
Catalog No. 502750945
Encompass_Preferred
The KISSLER Universal Fit Bolts are made from Grade 5 steel with zinc plating for corrosion resistance. They feature hex heads and are available in 3/8"-16 thread size in lengths ranging from 1" to 5". The bolts have a tensile strength of 120,000 psi. They can be used for general purpose fastening applications. The bolts meet ASME B18.2.1 specifications. They are sold in packs of 25, 50 or 100 pieces.
Catalog No. 50-275-0945 Supplier Grainger Supplier No. 14U361
The KISSLER Universal Fit Bolts are made from Grade 5 steel with zinc plating for corrosion resistance. They feature hex heads and are available in 3/8"-16 thread size in lengths ranging from 1" to 5". The bolts have a tensile strength of 120,000 psi. They can be used for general purpose fastening applications. The bolts meet ASME B18.2.1 specifications. They are sold in packs of 25, 50 or 100 pieces.