Learn More
CPC Scientific Glp-Gln-Arg-Leu-Gly-Asn-Gln-Trp-Ala-Val-Gly-His-Leu-Met-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific BOMB001A
Encompass
Tetraddecapeptide that is originally isolated from amphibian skin and is biologically active in both the central nervous system and gastrointestinal tract. Bombina bombina and bombina variegata variegata, a potent stimulant of gastric acid secretion, is proposed to be strong biologically active in central nervous system, e.g. suppressing feeding in rats. Bombesin-like peptides have been biochemically characterized in rat brain.SEQUENCE: Glp-Gln-Arg-Leu-Gly-Asn-Gln-Trp-Ala-Val-Gly-His-Leu-Met-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Glp-QRLGNQWAVGHLM-NH2MOLECULAR FORMULA: C71H110N24O18S1MOLECULAR WEIGHT: 1619.9STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [31362-50-2]SYNONYMS: RESEARCH AREA: ObesityREFERENCES:A. Anastasi et al., Experientia, 27, 166 (1971)SKU(s): Bomb-001A, Bomb-001B
Catalog No. 50-210-8841