Learn More
American Radiolabeled Chemicals Inc BOMBESIN 125I 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARI 017750UC
Encompass
Bombesin, Tyr-125I 50uCi in buffered solution (pH 3.4), 1 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~1794, Shipped in Dry Ice
Catalog No. 50-901-175
Add to Cart
Request a Quote