Learn More
Bachem Bombesin Trifluoroacetate
SDP
Supplier:  Bachem 4010579.0005
Encompass_Preferred
Bombesin Trifluoroacetate, Pyr-Gln-Arg-Leu-Gly-Asn-Gln-Trp-Ala-Val-Gly-His-Leu-Met-NH2, 31362-50-2, C71H110N24O18S, Mr 1619.87, 5mg. A tetradecapeptide isolated from amphibian skin, which is biologically active in both the central nervous system and gastrointestinal tract. It was also shown that bombesin suppresses feeding in rats. Bombesin-like peptides have been biochemically characterized in rat brain.
Catalog No. 50-259-573