Learn More
Medchemexpress LLC Bomedemstat dihydrochloride | 99.55% | 592.54 | 25 MG
SDP
Catalog No. 5000347519
Encompass_Preferred
Bomedemstat dihydrochloride is an orally active and irreversible lysine-specific demethylase 1 (LSD1) inhibitor. It increases H3K4 and H3K9 methylation, which then alters gene expression. This compound exhibits anti-cancer activities by inhibiting cancer cell proliferation and inducing apoptosis, and is being investigated in multiple clinical trials.
  • Selectively inhibits proliferation of Jak2V617F cells
  • Induces apoptosis via BCL-XL and PUMA in a TP53-dependent manner
  • Leads to cell cycle arrest in SET-2 cells
  • Normalizes or improves blood cell counts in Mx-Jak2V617F mice
  • Reduces spleen volumes and restores normal splenic architecture in animal models
Catalog No. 50-003-47519 Supplier Medchemexpress LLC Supplier No. 25MGHY109169C
Bomedemstat dihydrochloride is an orally active and irreversible lysine-specific demethylase 1 (LSD1) inhibitor. It increases H3K4 and H3K9 methylation, which then alters gene expression. This compound exhibits anti-cancer activities by inhibiting cancer cell proliferation and inducing apoptosis, and is being investigated in multiple clinical trials.
  • Selectively inhibits proliferation of Jak2V617F cells
  • Induces apoptosis via BCL-XL and PUMA in a TP53-dependent manner
  • Leads to cell cycle arrest in SET-2 cells
  • Normalizes or improves blood cell counts in Mx-Jak2V617F mice
  • Reduces spleen volumes and restores normal splenic architecture in animal models